The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285947.1 | NP_001272876.1 | 1 - 2 | 425 | 86 |
NM_001285948.1 | NP_001272877.1 | 1 - 2 | 425 | 86 | |
NM_018887.4 | NP_061375.1 | 1 - 2 | 425 | 86 | |
GenBank mRNA | AF237981.1 | - | 1 - 2 | 274 | 86 |
AK031733.1 | - | 1 - 2 | 373 | 86 | |
BC021418.1 | - | 1 - 2 | 296 | 86 | |
BC037663.1 | - | 1 - 2 | 274 | 86 | |
BE648427.1 | - | 1 - 2 | 86 |