The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019578.2 | NP_062524.2 | 9 - 10 | 2491 | 55 |
GenBank mRNA | AF224461.1 | - | 9 - 10 | 1723 | 55 |
AK085075.1 | - | 9 - 10 | 2493 | 55 | |
AK161915.1 | - | 9 - 10 | 1564 | 55 | |
AK172619.1 | - | 9 - 10 | 2493 | 55 | |
BC120890.1 | - | 9 - 10 | 1723 | 55 | |
BC120891.1 | - | 9 - 10 | 1723 | 55 |