The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025325.2 | NP_079601.1 | 1 - 2 | 152 | 65 |
GenBank mRNA | AK002295.1 | - | 1 - 2 | 174 | 65 |
AK002608.1 | - | 1 - 2 | 112 | 65 | |
AK171228.1 | - | 2 - 3 | 1306 | 65 | |
BC012872.1 | - | 1 - 2 | 152 | 65 | |
BC094021.1 | - | 1 - 2 | 152 | 65 |