Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddx56
Entrez Gene ID
52513
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 56
Gene Aliases
2600001H07Rik, D11Ertd619e, Noh61
Chromosome Location
Chr.11: 6257545 - 6267729 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026538.3 NP_080814.1 12 - 13 1500 63
GenBank mRNA AK011136.1 - 12 - 13 1510 63
AK076047.1 - 12 - 13 1518 63
AK076975.1 - 12 - 13 1500 63
AK077514.1 - 12 - 13 1513 63
AK077953.1 - 7 - 8 747 63
AK145628.1 - 12 - 13 1541 63
AK159793.1 - 12 - 13 1504 63
AK169654.1 - 12 - 13 1515 63
BC018291.1 - 12 - 13 1494 63

View Full Product Details