The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030609.3 | NP_085112.1 | 1 - 1 | 554 | 67 |
GenBank mRNA | BC116820.1 | - | 1 - 1 | 529 | 67 |
BC116850.1 | - | 1 - 1 | 527 | 67 |