Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gjc3
Entrez Gene ID
118446
Gene Name
gap junction protein, gamma 3
Gene Aliases
Cx29, Gje1
Chromosome Location
Chr.5: 137953809 - 137962959 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080450.4 NP_536698.2 2 - 2 144 91
GenBank mRNA AK034037.1 - 2 - 2 109 91
AK045577.1 - 2 - 2 284 91
AK047368.1 - 2 - 2 145 91
AK147913.1 - 2 - 2 284 91
AY289210.1 - 1 - 1 49 91
AY293564.1 - 1 - 1 49 91
AY297108.1 - 1 - 1 49 91
BC125305.1 - 2 - 2 131 91

View Full Product Details