The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145365.3 | NP_663340.1 | 11 - 12 | 1478 | 57 |
GenBank mRNA | AF392874.1 | - | 9 - 10 | 1129 | 57 |
AK050186.1 | - | 9 - 10 | 1120 | 57 | |
AK077258.1 | - | 11 - 12 | 1479 | 57 | |
BC010786.1 | - | 9 - 10 | 1173 | 57 | |
BC028820.1 | - | 9 - 10 | 1150 | 57 |