The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145419.1 | NP_663394.1 | 1 - 2 | 165 | 61 |
GenBank mRNA | AK044665.1 | - | 1 - 2 | 118 | 61 |
AK145980.1 | - | 1 - 2 | 144 | 61 | |
BC016235.1 | - | 1 - 2 | 165 | 61 |