The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145427.2 | NP_663402.2 | 3 - 4 | 543 | 69 |
GenBank mRNA | AK078500.1 | - | 3 - 4 | 488 | 69 |
BC013607.1 | - | 3 - 4 | 449 | 69 |