The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145499.2 | NP_663474.2 | 7 - 8 | 1176 | 68 |
GenBank mRNA | AK050082.1 | - | 7 - 8 | 1177 | 68 |
AY227736.1 | - | 7 - 8 | 1160 | 68 | |
BC016494.1 | - | 7 - 8 | 1161 | 68 | |
BC022151.1 | - | 7 - 8 | 1154 | 68 | |
BC023894.1 | - | 7 - 8 | 1168 | 68 | |
BC025822.1 | - | 7 - 8 | 1178 | 68 | |
BC034831.1 | - | 7 - 8 | 1160 | 68 |