Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gatad2a
Entrez Gene ID
234366
Gene Name
GATA zinc finger domain containing 2A
Gene Aliases
1110066C11Rik, BC031407, C80248
Chromosome Location
Chr.8: 69907069 - 69996384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113346.1 NP_001106817.1 9 - 10 1587 84
NM_001286450.1 NP_001273379.1 9 - 10 1744 84
NM_145596.4 NP_663571.2 9 - 10 1738 84
GenBank mRNA AK089074.1 - 9 - 10 1720 84
AK160058.1 - 9 - 10 1593 84
BC019178.1 - 1 - 2 104 84
BC031407.1 - 2 - 3 355 84
BC038221.1 - 9 - 10 1485 84
CF726663.1 - 1 - 2 126 84

View Full Product Details