The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161406.1 | NP_001154878.1 | 11 - 12 | 1614 | 78 |
NM_145890.2 | NP_665897.2 | 9 - 10 | 1246 | 78 | |
GenBank mRNA | AF411211.1 | - | 9 - 10 | 1246 | 78 |
AF411212.1 | - | 11 - 12 | 1585 | 78 | |
AK076307.1 | - | 11 - 12 | 1600 | 78 | |
AK161048.1 | - | 11 - 12 | 1496 | 78 | |
BC013080.1 | - | 11 - 12 | 1554 | 78 | |
BC037233.1 | - | 11 - 12 | 1606 | 78 |