Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h7a
Entrez Gene ID
106205
Gene Name
zinc finger CCCH type containing 7 A
Gene Aliases
A430104C18Rik, AI447294, AW556219, HSPC055, Zc3h7, Zc3hdc7
Chromosome Location
Chr.16: 11136594 - 11176393 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145931.2 NP_666043.2 3 - 4 318 114
NR_027502.1 3 - 4 318 114
GenBank mRNA AK028438.1 - 3 - 4 243 115
AK040507.1 - 3 - 4 266 114
AK049834.1 - 3 - 4 253 114
AK052830.1 - 4 - 5 429 114
AK150952.1 - 3 - 4 237 114
AK161148.1 - 3 - 4 296 114
AK166876.1 - 3 - 4 266 114
AK167758.1 - 3 - 4 267 114
BC027330.1 - 3 - 4 214 114
BC029320.1 - 3 - 4 216 114

View Full Product Details