Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Madcam1
Entrez Gene ID
17123
Gene Name
mucosal vascular addressin cell adhesion molecule 1
Gene Aliases
AV211525
Chromosome Location
Chr.10: 79664395 - 79668541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013591.2 NP_038619.2 1 - 2 95 79
GenBank mRNA AK137028.1 - 1 - 2 96 79
AK156096.1 - 1 - 2 73 79
BC024372.1 - 1 - 2 88 79
BC115910.1 - 1 - 2 89 79
BC118056.1 - 1 - 2 89 79
L21203.1 - 1 - 2 92 79
S80258.1 - 1 - 2 62 79

View Full Product Details