Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lpin1
Entrez Gene ID
14245
Gene Name
lipin 1
Gene Aliases
4631420P06, Kiaa0188, Lipin1, fld, mKIAA0188
Chromosome Location
Chr.12: 16535669 - 16646968 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130412.1 NP_001123884.1 13 - 14 1909 73
NM_015763.4 NP_056578.2 13 - 14 1864 73
NM_172950.3 NP_766538.2 12 - 13 1765 73
GenBank mRNA AB093214.1 - 8 - 9 1084 73
AF180471.1 - 12 - 13 1744 73
AF412811.1 - 12 - 13 1802 73
AK014526.1 - 12 - 13 1825 73
AK019539.1 - 13 - 14 2002 73
AK030921.1 - 12 - 13 1768 73
AK154837.1 - 13 - 14 1906 73
BC042462.1 - 12 - 13 1818 73

View Full Product Details