Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Luzp1
Entrez Gene ID
269593
Gene Name
leucine zipper protein 1
Gene Aliases
2700072H04Rik, AI266952, Luzp, mFLJ00226
Chromosome Location
Chr.4: 136469749 - 136554725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024452.2 NP_077772.2 3 - 4 284 94
GenBank mRNA AK139583.1 - 3 - 4 285 94
L49344.1 - 2 - 3 145 94

View Full Product Details