Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Luc7l
Entrez Gene ID
66978
Gene Name
Luc7 homolog (S. cerevisiae)-like
Gene Aliases
1810045C04Rik, 2410018D03Rik
Chromosome Location
Chr.17: 26245364 - 26285510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025881.3 NP_080157.1 7 - 8 901 73
NM_028190.3 NP_082466.2 7 - 8 901 73
NR_037905.1 8 - 9 972 73
NR_037906.1 7 - 8 1958 73
GenBank mRNA AK010539.1 - 7 - 8 853 73
AK017655.1 - 7 - 8 861 73
BC019194.1 - 8 - 9 894 73
BC034135.1 - 6 - 7 1067 73
BC055875.1 - 7 - 8 1884 73
BQ572015.1 - 5 - 6 553 73
CF951283.1 - 3 - 4 297 73

View Full Product Details