The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144785.2 | NP_659034.1 | 2 - 3 | 622 | 90 |
GenBank mRNA | AF529221.1 | - | 2 - 3 | 551 | 90 |
AK052752.1 | - | 2 - 3 | 592 | 90 | |
BC021384.1 | - | 2 - 3 | 588 | 90 |