The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144791.2 | NP_659040.2 | 6 - 7 | 1026 | 92 |
GenBank mRNA | BC010841.1 | - | 6 - 7 | 841 | 92 |