The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144838.1 | NP_659087.1 | 10 - 11 | 1001 | 63 |
GenBank mRNA | AK049820.1 | - | 10 - 11 | 1039 | 63 |
AK082080.1 | - | 10 - 11 | 891 | 63 | |
BC017611.1 | - | 10 - 11 | 1001 | 63 |