The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146125.2 | NP_666237.1 | 6 - 7 | 1215 | 73 |
GenBank mRNA | BC027291.1 | - | 6 - 7 | 1215 | 73 |