Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tspan33
Entrez Gene ID
232670
Gene Name
tetraspanin 33
Gene Aliases
1300010A20Rik, Pen
Chromosome Location
Chr.6: 29694214 - 29718558 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301407.1 NP_001288336.1 7 - 8 977 109
NM_146173.3 NP_666285.1 7 - 8 980 109
GenBank mRNA AF276890.1 - 7 - 8 979 109
AK004954.1 - 7 - 8 953 109
AK172176.1 - 7 - 8 958 109
BC024685.1 - 7 - 8 938 109

View Full Product Details