Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfp46
Entrez Gene ID
22704
Gene Name
zinc finger protein 46
Gene Aliases
AI426962, Mlz-4, Zfp-46, Znf436
Chromosome Location
Chr.4: 136284168 - 136293942 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009557.3 NP_033583.2 3 - 3 3904 81
GenBank mRNA AK148255.1 - 3 - 3 3855 81
AK173240.1 - 3 - 3 3916 81
BC092398.1 - 1 - 1 3214 81
M98502.1 - 1 - 1 3448 81

View Full Product Details