The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025665.2 | NP_079941.1 | 1 - 2 | 47 | 89 |
GenBank mRNA | AK011937.1 | - | 1 - 2 | 48 | 89 |
AK045231.1 | - | 1 - 2 | 60 | 89 | |
AK160190.1 | - | 1 - 2 | 57 | 89 | |
BC027564.1 | - | 1 - 2 | 48 | 89 |