Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc13
Entrez Gene ID
243983
Gene Name
zinc finger, DHHC domain containing 13
Gene Aliases
2410004E01Rik, C530010M18, HIP3RP, Hip14l, kojak, skc4
Chromosome Location
Chr.7: 48789003 - 48827440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028031.3 NP_082307.1 16 - 17 1836 78
GenBank mRNA AK010382.1 - 16 - 17 1836 78
AK145855.1 - 16 - 17 1756 78
AK146196.1 - 16 - 17 1808 78
BC019493.1 - 16 - 17 1755 78
BC046599.1 - 16 - 17 1755 78

View Full Product Details