Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hinfp
Entrez Gene ID
102423
Gene Name
histone H4 transcription factor
Gene Aliases
AA589481, HiNF-P, Mizf
Chromosome Location
Chr.9: 44292438 - 44305705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172162.3 NP_751894.2 3 - 4 462 73
GenBank mRNA AK049348.1 - 3 - 4 426 73
AK049633.1 - 3 - 4 623 73
BC029091.1 - 2 - 3 445 73
CN708951.1 - 2 - 3 308 73
CT010239.1 - 2 - 3 397 73

View Full Product Details