Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf40
Entrez Gene ID
233900
Gene Name
ring finger protein 40
Gene Aliases
AI848422, mKIAA0661
Chromosome Location
Chr.7: 127588698 - 127603605 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172281.2 NP_758485.2 8 - 9 1172 62
GenBank mRNA AK089805.1 - 8 - 9 1104 62
AK129184.1 - 8 - 9 1090 62
AK154982.1 - 8 - 9 1103 62
AK171462.1 - 8 - 9 1101 62
BC038348.1 - 8 - 9 1086 62

View Full Product Details