The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_180600.3 | NP_850931.2 | 6 - 7 | 951 | 125 |
GenBank mRNA | AK076148.1 | - | 6 - 7 | 814 | 125 |
AK083216.1 | - | 6 - 7 | 944 | 125 | |
AK150776.1 | - | 6 - 7 | 776 | 125 | |
BC029111.1 | - | 6 - 7 | 780 | 125 |