The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289613.1 | NP_001276542.1 | 1 - 2 | 708 | 77 |
NM_001289616.1 | NP_001276545.1 | 2 - 3 | 193 | 77 | |
NM_001289620.1 | NP_001276549.1 | 2 - 3 | 193 | 77 | |
NM_172434.3 | NP_766022.1 | 1 - 2 | 708 | 77 | |
GenBank mRNA | AK044069.1 | - | 1 - 2 | 141 | 77 |
AK044506.1 | - | 2 - 3 | 157 | 77 | |
AK087536.1 | - | 2 - 3 | 153 | 77 | |
AY165052.1 | - | 1 - 2 | 426 | 77 | |
BC057553.1 | - | 1 - 2 | 763 | 77 | |
BY089288.1 | - | 2 - 3 | 194 | 77 |