The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040426.3 | NP_001035516.2 | 8 - 9 | 1632 | 76 |
NM_172444.4 | NP_766032.1 | 2 - 3 | 440 | 76 | |
GenBank mRNA | AK045414.1 | - | 2 - 3 | 356 | 76 |
AK138976.1 | - | 8 - 9 | 1633 | 76 | |
BC139329.1 | - | 2 - 3 | 333 | 76 | |
CV559893.1 | - | 2 - 3 | 440 | 76 |