The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025748.4 | NP_080024.3 | 2 - 3 | 560 | 122 |
GenBank mRNA | AK016938.1 | - | 2 - 3 | 123 | |
AK021291.1 | - | 2 - 3 | 248 | 122 | |
BC062195.1 | - | 2 - 3 | 278 | 122 |