The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027654.2 | NP_081930.1 | 9 - 10 | 1031 | 80 |
GenBank mRNA | AB047007.1 | - | 9 - 10 | 1081 | 80 |
AK155067.1 | - | 9 - 10 | 1001 | 80 | |
BC016195.1 | - | 9 - 10 | 1031 | 80 | |
BC089460.1 | - | 9 - 10 | 1070 | 80 |