The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028355.3 | NP_082631.1 | 1 - 2 | 364 | 55 |
GenBank mRNA | AK013409.1 | - | 1 - 2 | 101 | 55 |
AK144734.1 | - | 1 - 2 | 365 | 55 | |
BC021337.1 | - | 1 - 2 | 119 | 55 | |
DQ141695.1 | - | 1 - 2 | 59 | 55 |