The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028880.3 | NP_083156.2 | 1 - 2 | 511 | 92 |
GenBank mRNA | AK053345.1 | - | 1 - 2 | 501 | 92 |
AK080836.1 | - | 1 - 2 | 542 | 91 | |
BC027803.1 | - | 1 - 2 | 393 | 92 |