Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcor
Entrez Gene ID
71458
Gene Name
BCL6 interacting corepressor
Gene Aliases
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575
Chromosome Location
Chr.X: 12036737 - 12160355 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168321.1 NP_001161793.1 10 - 11 4555 74
NM_029510.3 NP_083786.2 11 - 12 5380 74
NM_175044.3 NP_778209.2 10 - 11 5326 74
NM_175045.3 NP_778210.2 11 - 12 5278 74
NM_175046.3 NP_778211.2 10 - 11 5224 74
GenBank mRNA AK018034.1 - 1 - 2 146 74
AK018370.1 - 3 - 4 635 74
AK031282.1 - 3 - 4 682 74
AK129398.1 - 10 - 11 4555 74
AY161170.1 - 11 - 12 4695 74
AY161171.1 - 10 - 11 4641 74
AY161172.1 - 11 - 12 4593 74
AY161173.1 - 10 - 11 4539 74
BC038637.1 - 3 - 4 585 74
BC058656.1 - 11 - 12 4779 74

View Full Product Details