Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm1b
Entrez Gene ID
218214
Gene Name
lysine (K)-specific demethylase 1B
Gene Aliases
4632428N09Rik, AI482520, Aof1
Chromosome Location
Chr.13: 47043471 - 47085279 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172262.3 NP_758466.1 13 - 14 1732 72
GenBank mRNA BC023917.1 - 13 - 14 1673 72

View Full Product Details