Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps18
Entrez Gene ID
228545
Gene Name
vacuolar protein sorting 18 (yeast)
Gene Aliases
9930024E13Rik
Chromosome Location
Chr.2: 119288742 - 119298453 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172269.3 NP_758473.3 1 - 2 388 96
GenBank mRNA AK029109.1 - 1 - 2 365 96
AK033333.1 - 1 - 2 373 96
AK036915.1 - 1 - 2 373 96
AK039848.1 - 1 - 2 373 96
AK044821.1 - 1 - 2 391 96
AK148077.1 - 1 - 2 377 96
AK160689.1 - 1 - 2 365 96
BC036129.1 - 1 - 2 316 96
BC039176.1 - 1 - 2 295 96

View Full Product Details