The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172276.3 | NP_758480.2 | 6 - 7 | 1093 | 66 |
GenBank mRNA | AK031872.1 | - | 6 - 7 | 1084 | 66 |
AK088182.1 | - | 6 - 7 | 1037 | 66 | |
AK140369.1 | - | 6 - 7 | 1086 | 66 | |
BC023268.1 | - | 1 - 2 | 124 | 66 | |
BC057314.1 | - | 6 - 7 | 3023 | 66 |