The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172436.3 | NP_766024.1 | 5 - 6 | 493 | 70 |
GenBank mRNA | AK019150.1 | - | 5 - 6 | 495 | 70 |
AK030595.1 | - | 5 - 6 | 523 | 70 | |
AK083156.1 | - | 5 - 6 | 494 | 70 | |
AK147948.1 | - | 5 - 6 | 508 | 70 | |
BC024952.1 | - | 5 - 6 | 688 | 70 | |
BC060505.1 | - | 5 - 6 | 480 | 70 |