Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Inpp5j
Entrez Gene ID
170835
Gene Name
inositol polyphosphate 5-phosphatase J
Gene Aliases
Pib5pa, Pipp
Chromosome Location
Chr.11: 3494272 - 3505567 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172439.3 NP_766027.2 2 - 3 1338 69
GenBank mRNA AK034272.1 - 2 - 3 1339 69
AK136908.1 - 2 - 3 1325 69
AK165718.1 - 2 - 3 1709 69
AK170114.1 - 2 - 3 1304 69
AK171029.1 - 2 - 3 1299 69
BC022915.1 - 2 - 3 1291 69
BC046561.1 - 1 - 2 151 69
BC051452.1 - 2 - 3 1308 69
BC131634.1 - 2 - 3 1333 69
BC131635.1 - 2 - 3 1330 69

View Full Product Details