The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172632.2 | NP_766220.2 | 3 - 4 | 1641 | 135 |
GenBank mRNA | AK079654.1 | - | 3 - 4 | 1619 | 135 |
BC058942.1 | - | 2 - 3 | 1044 | 135 | |
BC062911.1 | - | 2 - 3 | 1044 | 135 |