Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rsbn1
- Entrez Gene ID
-
229675
- Gene Name
- rosbin, round spermatid basic protein 1
- Gene Aliases
-
C230004D03Rik, E330004N01, Rosbin, Rsbn, Rsbp, mKIAA3002
- Chromosome Location
-
Chr.3: 103914067 - 103966620 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB101658.1 |
- |
7 - 8 |
2721 |
87 |
AK087685.1 |
- |
6 - 7 |
1685 |
87 |
View Full Product Details