Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fgd5
Entrez Gene ID
232237
Gene Name
FYVE, RhoGEF and PH domain containing 5
Gene Aliases
C330025N11Rik, ZFYVE23, mFLJ00274
Chromosome Location
Chr.6: 91978821 - 92076005 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172731.2 NP_766319.2 18 - 19 4031 80
GenBank mRNA AK049341.1 - 5 - 6 705 80
AK086031.1 - 4 - 5 541 80
AK220153.1 - 19 - 20 4505 80
BC042732.1 - 14 - 15 1448 80
BC060664.1 - 18 - 19 3494 80
BC145502.1 - 19 - 20 3848 80
BC150786.1 - 18 - 19 3980 80

View Full Product Details