Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csgalnact1
Entrez Gene ID
234356
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
4732435N03Rik, CSGalNAcT-1
Chromosome Location
Chr.8: 68356781 - 68735673 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252623.1 NP_001239552.1 10 - 11 2313 65
NM_172753.5 NP_766341.4 9 - 10 2009 65
GenBank mRNA AK028688.1 - 9 - 10 1932 65
AK033522.1 - 7 - 8 1663 65
AK080687.1 - 9 - 10 1978 65
AK133567.1 - 7 - 8 1662 65
AK144089.1 - 6 - 7 1570 65
BC026599.1 - 4 - 5 429 65
BC057630.1 - 11 - 12 2281 65
BC089162.1 - 9 - 10 2006 65

View Full Product Details