The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285956.1 | NP_001272885.1 | 10 - 11 | 1998 | 63 |
NM_001285958.1 | NP_001272887.1 | 10 - 11 | 1894 | 63 | |
NM_172874.3 | NP_766462.2 | 10 - 11 | 1920 | 63 | |
GenBank mRNA | AK034987.1 | - | 9 - 10 | 2451 | 63 |
AK161018.1 | - | 10 - 11 | 1784 | 63 | |
AY313606.1 | - | 10 - 11 | 1920 | 63 | |
BC052487.1 | - | 10 - 11 | 1745 | 63 | |
BC094340.1 | - | 10 - 11 | 1998 | 63 |