The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172904.2 | NP_766492.2 | 11 - 12 | 1901 | 66 |
GenBank mRNA | AK036674.1 | - | - | 1867 | 66 |
AK052866.1 | - | 5 - 6 | 701 | 66 |