The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173007.3 | NP_766595.1 | 1 - 2 | 114 | 88 |
GenBank mRNA | AK050737.1 | - | 1 - 2 | 115 | 88 |