Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smg5
Entrez Gene ID
229512
Gene Name
Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
BC024683, mKIAA1089
Chromosome Location
Chr.3: 88336260 - 88362338 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178246.3 NP_839977.2 6 - 7 797 94
GenBank mRNA AK129286.1 - 6 - 7 776 94
AK155787.1 - 6 - 7 786 94
BC024683.1 - 6 - 7 749 94

View Full Product Details