Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mb21d1
Entrez Gene ID
214763
Gene Name
Mab-21 domain containing 1
Gene Aliases
E330016A19Rik, cGAS
Chromosome Location
Chr.9: 78430518 - 78443524 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK030292.1 - 4 - 5 1340 78

View Full Product Details