The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173403.2 | NP_775579.2 | 1 - 2 | 752 | 67 |
GenBank mRNA | AK087479.1 | - | 1 - 2 | 753 | 67 |
AK149202.1 | - | - | 753 | 67 | |
BC132529.1 | - | 1 - 2 | 664 | 67 | |
BC138048.1 | - | 1 - 2 | 664 | 67 |